Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASB7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ASB7 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15759920
![]() |
Novus Biologicals
NBP15759920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157599
![]() |
Novus Biologicals
NBP157599 |
100 μL |
Each for $487.50
|
|
|||||
Description
ASB7 Polyclonal specifically detects ASB7 in Human samples. It is validated for Western Blot.Specifications
ASB7 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
ankyrin repeat and SOCS box containing 7, ankyrin repeat and SOCS box protein 7, ankyrin repeat and SOCS box-containing 7, ASB-7, FLJ22551, FLJ25192 | |
ASB7 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q9H672 | |
140460 | |
Synthetic peptides corresponding to ASB7 (ankyrin repeat and SOCS box-containing 7) The peptide sequence was selected from the middle region of ASB7)(50ug). Peptide sequence QTPLHLSALRDDVLCARMLYNYGADTNTRNYEGQTPLAVSISISGSSRPC. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title