Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASB8 Antibody (OTI3A12), FITC, Novus Biologicals™
SDP

Catalog No. NBP272400F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272400F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272400F Supplier Novus Biologicals Supplier No. NBP272400F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ASB8 Monoclonal specifically detects ASB8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen ASB8
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3A12
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ankyrin repeat and SOCS box containing 8, ankyrin repeat and SOCS box protein 8, ankyrin repeat and SOCS box-containing 8, ASB-8, FLJ21255, MGC5540
Gene Symbols ASB8
Host Species Mouse
Immunogen Full length recombinant protein of human ASB8 (NP_077000) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 140461.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.