Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASB9 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB319762 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB319762 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB319762 Supplier Novus Biologicals Supplier No. NBP298662PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASB9 Polyclonal antibody specifically detects ASB9 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ASB9
Applications Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ankyrin repeat and SOCS box containing 9, ankyrin repeat and SOCS box protein 9, ankyrin repeat and SOCS box-containing 9, ankyrin repeat and suppressor of cytokine signaling box protein 9, ASB-9, DKFZP564L0862, FLJ20636, MGC4954
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ASB9 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 140462
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.