Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASC/TMS1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB218052 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB218052 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB218052 Supplier Novus Biologicals Supplier No. NBP177297AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASC/TMS1 Polyclonal antibody specifically detects ASC/TMS1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ASC/TMS1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias apoptosis-associated speck-like protein containing a CARD, ASC, CARD5, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYCARD, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS, TMS1, TMS-1
Host Species Rabbit
Immunogen This ASC/TMS1 Antibody was developed against a 14 amino acid peptide near the carboxy terminus of human ASC/TMS1 . The immunogen is located within the last 50 amino acids of ASC. Amino Acid Sequence: RESQSYLVEDLERS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 29108
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.