Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASC/TMS1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB284095 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB284095 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB284095 Supplier Novus Biologicals Supplier No. NBP178977MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASC/TMS1 Polyclonal antibody specifically detects ASC/TMS1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Flow (Intracellular)

Specifications

Antigen ASC/TMS1
Applications Flow Cytometry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, Flow (Intracellular)
Formulation 50mM Sodium Borate
Gene Alias apoptosis-associated speck-like protein containing a CARD, ASC, CARD5, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYCARD, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS, TMS1, TMS-1
Host Species Rabbit
Immunogen This ASC/TMS1 Antibody was developed against a synthetic peptide made to an N-terminal portion of the human ASC/TMS1 protein (between residues 1-50) [Uniprot: Q9ULZ3]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 29108
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.