Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASC/TMS1 Antibody (OTI3E9), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP271853R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271853R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271853R Supplier Novus Biologicals Supplier No. NBP271853R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ASC/TMS1 Monoclonal specifically detects ASC/TMS1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen ASC/TMS1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI3E9
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD
Gene Symbols PYCARD
Host Species Mouse
Immunogen This ASC/TMS1 Antibody (OTI3E9) was developed against full length human recombinant protein of human ASC/TMS1 (NP_037390) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 29108.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.