Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASC1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB241150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB241150 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB241150 Supplier Novus Biologicals Supplier No. NB100419B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASC1 Polyclonal specifically detects ASC1 in Human samples. It is validated for Immunoprecipitation, Proximity Ligation Assay, Western Blot.

Specifications

Antigen ASC1
Applications Immunoprecipitation, Proximity Ligation Assay, Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias activating signal cointegrator 1, ASC-1, HsT17391, thyroid hormone receptor interactor 4, thyroid receptor interacting protein 4, Thyroid receptor-interacting protein 4, TR-interacting protein 4, TRIP-4
Host Species Rabbit
Immunogen A synthetic peptide mapping to a region between residues 75 and 125 of human ASC1 using the numbering given in entry NP_057297.2 (GeneID 9325.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9325
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.