Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASC2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056076T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056076T 0.025 mL
NB10056076 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056076T Supplier Novus Biologicals Supplier No. NB10056076SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

ASC2 Polyclonal specifically detects ASC2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen ASC2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Neat whole antisera with 0.05% Sodium Azide
Gene Alias ASC2PAAD-only protein 1, ASCI, POP1pyrin domain-containing protein 1, PYC1, PYD (pyrin domain) containing 1, pyrin domain containing 1, pyrin-domain containing protein 1, Pyrin-only protein 1
Gene Symbols PYDC1
Host Species Rabbit
Immunogen A recombinant full-length human ASC-2/POP1 protein was used as immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 260434
Test Specificity In immuhistochemical studies, ASC-2/POP1 has been found to expressed predominantly in macrophages and granulocytes (Stehlik et al, 2003).
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.