Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbate Oxidase Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB100200401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100200401 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100200401 Supplier Novus Biologicals Supplier No. NB10020040.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ascorbate Oxidase Polyclonal specifically detects Ascorbate Oxidase in Plant samples. It is validated for ELISA, Immunoprecipitation, Western Blot.

Specifications

Antigen Ascorbate Oxidase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 1:500-1:5000, ELISA 1:5000-1:20000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias Ascorbase, ASO, L ascorbate oxidase precursor
Gene Symbols AAO
Host Species Rabbit
Immunogen Ascorbate Oxidase [cucurbita]
Purification Method Delipidation and Defibrination
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Biotin, anti-Rabbit Serum as well as purified and partially purified Ascorbate Oxidase [cucurbita].
Target Species Plant
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.