Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbate Oxidase Antibody, Novus Biologicals™
SDP

Catalog No. NBP244233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244233 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244233 Supplier Novus Biologicals Supplier No. NBP244233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Ascorbate Oxidase in Plant samples, and it is validated for ELISA, Immunoprecipitation, Western Blotting

Ascorbate Oxidase Polyclonal specifically detects Ascorbate Oxidase in Plant samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Ascorbate Oxidase
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, ELISA 1:25000-1:125000, Immunoprecipitation 1:100
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P24792
Gene Alias Ascorbase, ASO, L ascorbate oxidase precursor
Gene Symbols AAO
Host Species Rabbit
Immunogen Ascorbate Oxidase [cucurbita] (Uniprot: P24792)
Molecular Weight of Antigen 64.67 kDa
Purification Method Multi-step
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Plant
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.