Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ascorbic Acid, Fine Powder, 200 to 325 Mesh, Multi-Compendial, U.S.P., J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Specifications
Mesh Size | 200 to 325 Mesh |
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 1 kg |
Specific Gravity | 1L = 1.65kg |
Formula Weight | 176.12 |
Grade | Multi-Compendial/USP |
Physical Form | Fine Powder |
Chemical Name or Material | Ascorbic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction