Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ascorbic Acid (Powder/USP/FCC), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
Specifications
Specifications
Melting Point | 190°C |
Color | Yellow |
Density | 1.65g/cm³ |
Ignition Residue | 0.1% max. (FCC, USP) |
Packaging | Poly Pail |
Linear Formula | CH2OHCHOHCHCOH:COHCOO |
Quantity | 12 kg |
Specific Rotation | +20.5 to +21.5° (FCC, USP) |
Residual Solvents | Meets Requirements (USP) |
Formula Weight | 176.13 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction