Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbic Acid, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18600778 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

AS102, 50-81-7, C6H8O6

Spectrum™ Chemical Ascorbic Acid, USP , Vitamin C, is a dietary supplement used to treat and prevent vitamin C deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 500 g
Specific Rotation +20.5° to +21.5°
Percent Purity 99 to 100.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.