Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASF1a Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB105699 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105699 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105699 Supplier Novus Biologicals Supplier No. NBP298775UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASF1a Polyclonal specifically detects ASF1a in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ASF1a
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias ASF1 silencing function 1 homolog A (S. cerevisiae), CCG1-interacting factor A, CIACGI-98, DKFZP547E2110, hAsf1, hAsf1a, hCIA, histone chaperone ASF1A, HSPC146, silencing function 1A, silencing function protein 1 homolog A
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ASF1a.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 25842
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.