Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASF1b Antibody (1D4), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP242676R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242676R 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242676R Supplier Novus Biologicals Supplier No. NBP242676R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ASF1b Monoclonal specifically detects ASF1b in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ASF1b
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1D4
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ASF1 silencing function 1 homolog B (S. cerevisiae), CCG1-interacting factor A-II, CIA-II, FLJ10604, hAsf1, hAsf1b, hCIA-II, histone chaperone ASF1B, silencing function protein 1 homolog B
Gene Symbols ASF1B
Host Species Mouse
Immunogen Recombinant human ASF1B (1-202aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 55723
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.