Learn More
Description
Specifications
Specifications
| Antigen | ASF1b |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1D4 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | ASF1 silencing function 1 homolog B (S. cerevisiae), CCG1-interacting factor A-II, CIA-II, FLJ10604, hAsf1, hAsf1b, hCIA-II, histone chaperone ASF1B, silencing function protein 1 homolog B |
| Gene Symbols | ASF1B |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
