Learn More
Description
Specifications
Specifications
| Antigen | ASGPR1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 103 |
| Conjugate | DyLight 488 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ASGPR1 (NP_033844.1; Ser 60-Asn 284). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
