Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASH1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N1093290A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1093290A30 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1093290A30 Supplier Novus Biologicals Supplier No. NB10093290AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASH1 Polyclonal specifically detects ASH1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation.

Specifications

Antigen ASH1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Absent small and homeotic disks protein 1 homolog, ASH1, ash1 (absent, small, or homeotic)-like (Drosophila), ASH1L1, ASH1-like protein, EC 2.1.1.43, FLJ10504, huASH1, KMT2HKIAA1420, Lysine N-methyltransferase 2H, probable histone-lysine N-methyltransferase ASH1L
Gene Symbols ASH1L
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2919 and 2969 of human absent, small and homeotic disks protein 1 using the numbering given in entry CAI12722.1 (GeneID 55870).
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 55870.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.