Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Asialoglycoprotein Receptor 2 Antibody (OTI2A12), FITC, Novus Biologicals™
SDP

Catalog No. NBP270212F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270212F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270212F Supplier Novus Biologicals Supplier No. NBP270212F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Asialoglycoprotein Receptor 2 Monoclonal specifically detects Asialoglycoprotein Receptor 2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Asialoglycoprotein Receptor 2
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI2A12
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ASGPR 2, ASGP-R 2, ASGPR2, ASGP-R2, asialoglycoprotein receptor 2, CLEC4H2FLJ60040, C-type lectin domain family 4 member H2, HBxAg-binding protein, HBXBP, Hepatic lectin H2, HL-2
Gene Symbols ASGR2
Host Species Mouse
Immunogen Full length recombinant protein of human Asialoglycoprotein Receptor 2 (NP_001172) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 433.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.