Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASIC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
$597.00
Specifications
Antigen | ASIC3 |
---|---|
Concentration | 0.20 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111401
|
Invitrogen™
PA5111401 |
100 μL |
Each of 1 for $597.00
|
|
|||||
Description
Immunogen sequence: CNINPLRRSR LTPNDLHWAG SALLGLDPAE HAAFLRALGR PPAPPGFMPS PTFDMAQLYA RAGHSLDDML LDCRFRGQPC GPENFTTIFT RM.
ACCN3 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, two hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. ACCN3 encodes a member that is an acid sensor and may play an important role in the detection of lasting pH changes. In addition, a heteromeric association between this member and ACCN1 has been observed as proton-gated channels sensitive to gadolinium.Specifications
ASIC3 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
ASIC3 | |
ACCN3; acid sensing (proton gated) ion channel 3; acid sensing ion channel 3; acid sensing ion channel subunit 3; acid-sensing (proton-gated) ion channel 3; acid-sensing ion channel 3; Amiloride-sensitive cation channel 3; amiloride-sensitive cation channel 3, testis; Asic3; AW742291; dorsal root ASIC; DRASIC; hASIC3; hTNaC1; modulatory subunit of ASIC2a; neuronal amiloride-sensitive cation channel 3; proton gated cation channel DRASIC; proton-gated cation channel subunit; SLNAC1; Testis sodium channel 1; TNaC1 | |
ASIC3 | |
Primary | |
Antigen affinity chromatography |
0.20 mg/mL | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UHC3 | |
9311 | |
Recombinant Protein corresonding to Human ASIC3. Recombinant protein control fragment (Product #RP-108208). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title