Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASIP Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093819 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093819 100 μL
NB093820 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093819 Supplier Novus Biologicals Supplier No. NBP304874100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASIP Polyclonal specifically detects ASIP in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ASIP
Applications Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias agouti (mouse)-signaling protein, agouti signaling protein, agouti signaling protein, nonagouti homolog, agouti signaling protein, nonagouti homolog (mouse), Agouti switch protein, agouti-signaling protein, AGTI, AGTILMGC126092, ASPAGSW, MGC126093, nonagouti homolog, SHEP9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 23-132 of human ASIP (NP_001663.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 434
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.