Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASK1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB357624 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357624 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357624 Supplier Novus Biologicals Supplier No. NBP176674MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASK1 Polyclonal antibody specifically detects ASK1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen ASK1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Apoptosis signal-regulating kinase 1, ASK-1, ASK1MEKK 5, EC 2.7.11, MAP/ERK kinase kinase 5, MAPK/ERK kinase kinase 5, MAPKKK5EC 2.7.11.25, MEK kinase 5, MEKK5apoptosis signal regulating kinase 1, mitogen-activated protein kinase kinase kinase 5
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ASK1. The immunogen is located within amino acids 1260 - 1310 of ASK1. Amino Acid Squence: IEEKDQEIKHLKLKSQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4217
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.