Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASK1, p Ser966 Antibody, Novus Biologicals™
SDP

Catalog No. NB176993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176993 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176993 Supplier Novus Biologicals Supplier No. NBP313306
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASK1 Polyclonal antibody specifically detects ASK1 in Human samples. It is validated for Western Blot

Specifications

Antigen ASK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS, 1% BSA, 20% Glycerol (pH7)
Gene Alias Apoptosis signal-regulating kinase 1, ASK-1, ASK1MEKK 5, EC 2.7.11, MAP/ERK kinase kinase 5, MAPK/ERK kinase kinase 5, MAPKKK5EC 2.7.11.25, MEK kinase 5, MEKK5apoptosis signal regulating kinase 1, mitogen-activated protein kinase kinase kinase 5
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide corresponding to residues around human ASK1 (phospho Ser966). The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4217
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.