Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASNA1 Antibody (2A1), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP242661AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242661AF 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242661AF Supplier Novus Biologicals Supplier No. NBP242661AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ASNA1 Monoclonal specifically detects ASNA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ASNA1
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2A1
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ARSA, arsA arsenite transporter, ATP-binding, homolog 1 (bacterial), ARSA1, ARSA-I, Arsenical pump-driving ATPase, Arsenite-stimulated ATPase, ATPase ASNA1, EC 3.6, EC 3.6.3.16, GET3, golgi to ER traffic 3 homolog, hARSA-I, hASNA-I, MGC3821, Transmembrane domain recognition complex 40 kDa ATPase subunit, transmembrane domain recognition complex, 40kDa, TRC40arsA (bacterial) arsenite transporter, ATP-binding, homolog 1
Gene Symbols GET3
Host Species Mouse
Immunogen Recombinant human ASNA1 (1-348aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 439
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.