Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASNA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ASNA1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ASNA1 Polyclonal specifically detects ASNA1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ASNA1 | |
Polyclonal | |
Rabbit | |
metabolism | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
439 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:EPTLSNIIEQRSLKWIFVGGKGGVGKTTCSCSLAVQLSKGRESVLIISTDPAHNISDAFDQKFSKVPTKVKGYDNLFAMEIDPS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
ARSA, arsA arsenite transporter, ATP-binding, homolog 1 (bacterial), ARSA1, ARSA-I, Arsenical pump-driving ATPase, Arsenite-stimulated ATPase, ATPase ASNA1, EC 3.6, EC 3.6.3.16, GET3, golgi to ER traffic 3 homolog, hARSA-I, hASNA-I, MGC3821, Transmembrane domain recognition complex 40 kDa ATPase subunit, transmembrane domain recognition complex, 40kDa, TRC40arsA (bacterial) arsenite transporter, ATP-binding, homolog 1 | |
GET3 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title