Learn More
Asparagine synthetase Antibody (OTI1A10), Alexa Fluor™ 405, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Asparagine synthetase |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI1A10 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | asparagine synthetase, asparagine synthetase (glutamine-hydrolyzing), asparagine synthetase [glutamine-hydrolyzing], Cell cycle control protein TS11, EC 6.3.5.4, Glutamine-dependent asparagine synthetase, TS11, TS11 cell cycle control protein |
| Gene Symbols | ASNS |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.