Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Asparagine synthetase Antibody (OTI1A10), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289747 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289747 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289747 Supplier Novus Biologicals Supplier No. NBP270215APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Asparagine synthetase Monoclonal antibody specifically detects Asparagine synthetase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Asparagine synthetase
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1A10
Conjugate APC
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias asparagine synthetase, asparagine synthetase (glutamine-hydrolyzing), asparagine synthetase [glutamine-hydrolyzing], Cell cycle control protein TS11, EC 6.3.5.4, Glutamine-dependent asparagine synthetase, TS11, TS11 cell cycle control protein
Host Species Mouse
Immunogen Full length human recombinant protein of human ASNS(NP_597680) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 440
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.