Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aspartate Aminotransferase Antibody (SR1790), Novus Biologicals™
SDP

Catalog No. NB259923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259923 50 μg
NB259759 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259923 Supplier Novus Biologicals Supplier No. NBP32184250UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Aspartate Aminotransferase Monoclonal antibody specifically detects Aspartate Aminotransferase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Aspartate Aminotransferase
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR1790
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias aspartate aminotransferase, cytoplasmic, ASTQTL1, CASPAT, CCAT, Cysteine aminotransferase, cytoplasmic, Cysteine transaminase, cytoplasmic, EC 2.6.1.1, EC 2.6.1.3, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), GOT1, growth-inhibiting protein 18, Transaminase A
Host Species Rabbit
Immunogen A synthesized peptide derived from human Aspartate Aminotransferase (Uniprot #: P17174)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2805
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.