Learn More
Description
Specifications
Specifications
| Antigen | Aspartate beta hydroxylase |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | AAH, ASP beta-hydroxylase, aspartate beta-hydroxylaseA beta H-J-J, aspartyl/asparaginyl-beta-hydroxylase, BAH, cardiac junctin, CASQ2BP1, EC 1.14.11.16, HAAHaspartyl/asparaginyl beta-hydroxylase, humbug, JCTN, junctate, junctin, Peptide-aspartate beta-dioxygenase |
| Gene Symbols | ASPH |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
