Learn More
Aspartate Transaminase Polyclonal Antibody, Rockland™

Sheep Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | Aspartate Transaminase |
---|---|
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Lyophilized |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1245
![]() ![]() |
Rockland Immunochemicals
2006011440100 |
100 μg |
Each for $636.24
|
|
|||||
50-199-1246
![]() ![]() |
Rockland Immunochemicals
200601144S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Aspartate Transaminase is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Sheep Serum as well as purified and partially purified Aspartate Transaminase (AST) [Pig Heart]. Cross reactivity against Aspartate Transaminase (AST) from other tissues and species may occur but have not been specifically determined.
GOT1 is a glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.Specifications
Aspartate Transaminase | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
AI789014; aspartate aminotransferase 1; Aspartate aminotransferase, cytoplasmic; aspartate transaminase 1; Aspat; AST1; ASTQTL1; cAspAT; cCAT; Cysteine aminotransferase, cytoplasmic; Cysteine transaminase, cytoplasmic; cytosolic aspartate aminotransferase; Gaspat; GIG18; Glutamate oxaloacetate transaminase 1; glutamate oxaloacetate transaminase 1, soluble; glutamic-oxaloacetic transaminase 1; glutamic-oxaloacetic transaminase 1, soluble; Got1; Got-1; growth-inhibiting protein 18; testis secretory sperm-binding protein Li 196a; Transaminase A | |
GOT1 | |
Primary | |
Antibody |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Sheep | |
Pig | |
P00503 | |
396967 | |
Anti-Aspartate Transaminase antibody was produced by repeated immunizations with Aspartate Transaminase from pig heart. | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
GOT1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.