Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aspergillus Antibody (343/31), Novus Biologicals™
SDP

Catalog No. NB10064511 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064511 0.1 mg
NB017591 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064511 Supplier Novus Biologicals Supplier No. NB10064511
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aspergillus Monoclonal specifically detects Aspergillus in Fungi samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen Aspergillus
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 343/31
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Alias Aspergillus spp
Host Species Mouse
Immunogen Native Aspergillus
Purification Method IgM purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64511 is specific for Aspergillus spp.
Target Species Fungi
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.