Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASPM Antibody, Novus Biologicals™
SDP

Catalog No. NB1002278 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002278 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002278 Supplier Novus Biologicals Supplier No. NB1002278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

ASPM Polyclonal specifically detects ASPM in Bovine, Human, Mouse, Ovine, Pig samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen ASPM
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q8IZT6
Gene Alias ASP, asp (abnormal spindle) homolog, microcephaly associated (Drosophila), Asp homolog, Calmbp1, FLJ43117, MCPH5DKFZp686N06184, microcephaly associated (Drosophila), microcephaly, primary autosomal recessive 5
Gene Symbols ASPM
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 3425 and the C-terminus (residue 3477) of human ASP (Abnormal Spindle)-like, Microcephaly Associated using the numbering given in entry NP_060606.2 (GeneID 259266).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259266
Target Species Bovine, Human, Mouse, Ovine, Pig
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.