Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Asprosin Antibody (Birdy-1), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB120679 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120679 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120679 Supplier Novus Biologicals Supplier No. NBP311677JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Asprosin Monoclonal specifically detects Asprosin in Human, Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Asprosin
Applications Western Blot, ELISA
Classification Monoclonal
Clone Birdy-1
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias FBN1, fibrillin 1
Host Species Mouse
Immunogen Recombinant human asprosin.
Purification Method >95%, by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2200
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.