Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASPRV1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen ASPRV1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP156511
SDP
View Documents
Novus Biologicals
NBP156511
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

ASPRV1 Polyclonal specifically detects ASPRV1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ASPRV1
Polyclonal
Rabbit
Q53RT3
151516
Synthetic peptides corresponding to SASP The peptide sequence was selected from the middle region of SASP. Peptide sequence RGEALGVYNRLSPQDQGDYGTVKEALLKAFGVPGAAPSHLPKEIVFANSM.
Primary
Western Blot
Unconjugated
RUO
aspartic peptidase, retroviral-like 1, EC 3.4.23.-, FLJ25084, retroviral-like aspartic protease 1, SASPaseMUNO, SASPTAPS, Skin aspartic protease, Skin-specific retroviral-like aspartic protease, Taps, TPA-inducible aspartic proteinase-like protein
ASPRV1
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.