Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASRGL1 Antibody (CRASH/1289), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP2440A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2440A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2440A70 Supplier Novus Biologicals Supplier No. NBP254450AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ASRGL1 Monoclonal specifically detects ASRGL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ASRGL1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CRASH/1289
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase
Gene Symbols ASRGL1
Host Species Mouse
Immunogen Recombinant human ASRGL1 fragment (Uniprot: Q7L266)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 80150.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.