Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protective Industrial Products Assurance™ 20 Mil Butyl Rubber Glove with Smooth Grip - 14 in.
Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- 20 Mil Bromine butyl rubber
- The addition of bromine enhances temperature resistance, offering superior durability
- 14 in. length for extended forearm protection
- Smooth grip
- Protects against hazardous chemicals and materials
- Easy to decontaminate
- Excellent holdout
- Maintains flexibility from -40°C to +90°C (-40°F to +194°F)
- ESD safe 10^8 Ohms
Specifications
Specifications
Applications | Laboratory |
Color | Black |
Latex Free | Yes |
Material | Butyl |
Product Type | Butyl Gloves |
Size | Medium |
Thickness (English) | 20 mil |
Thickness (Metric) | 0.508 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction