Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protective Industrial Products Assurance™ 20 Mil Butyl Rubber Glove with Smooth Grip - 14 in.

Catalog No. 18813340 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large

Catalog No. 18-813-340

Supplier: Protective Industrial Products BUT05/11

Only null left
Add to Cart
Add to Cart
Request a Quote

Designed to handle extreme exposure to polar hydrocarbons such as ketones, acids, esters, and amine derivatives.

  • 20 Mil Bromine butyl rubber
  • The addition of bromine enhances temperature resistance, offering superior durability
  • 14 in. length for extended forearm protection
  • Smooth grip
  • Protects against hazardous chemicals and materials
  • Easy to decontaminate
  • Excellent holdout
  • Maintains flexibility from -40°C to +90°C (-40°F to +194°F)
  • ESD safe 10^8 Ohms

Specifications

Applications Laboratory
Color Black
Latex Free Yes
Material Butyl
Product Type Butyl Gloves
Size 2X-Large
Thickness (English) 20 mil
Thickness (Metric) 0.508 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.