Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASTE1 Antibody, Novus Biologicals™
SDP

Catalog No. NB8029120UL Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB8029120UL 20 μL
NBP180291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB8029120UL Supplier Novus Biologicals Supplier No. NBP18029120UL

Rabbit Polyclonal Antibody

ASTE1 Polyclonal specifically detects ASTE1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen ASTE1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_079927
Gene Alias asteroid homolog 1 (Drosophila), HT001, MGC129980, protein asteroid homolog 1
Gene Symbols ASTE1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human Aste1. Peptide sequence YHRVLGLLNWLSHFDDPTEALDNVLKSLPKKSRENVKELLCCSMEEYQQS.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28990
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.