Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASTN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159195
Description
ASTN2 Polyclonal specifically detects ASTN2 in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
| ASTN2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| astrotactin 2, bA264C15.1, bA67K19.1, KIAA0634astrotactin-2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23245 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Q5JVY1 | |
| ASTN2 | |
| Synthetic peptides corresponding to ASTN2(astrotactin 2) The peptide sequence was selected from the N terminal of ASTN2. Peptide sequence LLFVRNELPGRIAVQDDLDNTELPFFTLEMSGTAADISLVHWRQQWLENG The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Guinea pig: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%. | |
| Human, Mouse, Rat, Guinea Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction