Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASTN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $501.50
Specifications
| Antigen | ASTN2 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15919520
![]() |
Novus Biologicals
NBP15919520UL |
20 μL |
Each for $207.00
|
|
|||||
NBP159195
![]() |
Novus Biologicals
NBP159195 |
100 μL |
Each for $501.50
|
|
|||||
Description
ASTN2 Polyclonal specifically detects ASTN2 in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
| ASTN2 | |
| Polyclonal | |
| Rabbit | |
| Q5JVY1 | |
| 23245 | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| astrotactin 2, bA264C15.1, bA67K19.1, KIAA0634astrotactin-2 | |
| ASTN2 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title