Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ASTN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597478
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597478 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597478 Supplier Invitrogen™ Supplier No. PA597478
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ASTN2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene encodes a protein that is expressed in the brain and may function in neuronal migration, based on functional studies of the related astrotactin 1 gene in human and mouse. A deletion at this locus has been associated with schizophrenia. Multiple transcript variants encoding different proteins have been found for this locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASTN2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ASTN2
Gene Accession No. O75129
Gene Alias 1d8; Astn2; Astnl; astrotactin 2; astrotactin 2-like; astrotactin-2; bA264C15.1; bA67K19.1; bM243L4.1 (astrotactin 2); bM264P11.1 (astrotactin 2); bM383P24.1 (astrotactin 2); bM423P8.1 (astrotactin 2); bM452J22.1; bM452J22.1 (novel protein); bM468K2.1 (astrotactin 2); Kiaa0634
Gene Symbols ASTN2
Host Species Rabbit
Immunogen Recombinant Human Astrotactin-2 protein (108-272AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23245
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.