Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Astrocytomas Antibody (J1-31), Novus Biologicals™
SDP

Catalog No. NBP229820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229820 0.1 mL
NB019477 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229820 Supplier Novus Biologicals Supplier No. NBP229820
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Astrocytomas Monoclonal specifically detects Astrocytomas in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Astrocytomas
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone J1-31
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Ascites with No Preservative
Gene Alias Astrocytoma Marker, Astrocytomas Marker, Low Grade Astrocytoma Marker
Host Species Mouse
Immunogen Human cerebral white matter plaque materials from a multiple sclerosis patient.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody recognizes an intracellular protein antigen (MW 30 kDa) expressed by human and rat astrocytes and other specialized glia (Muller cells of the retina, Bergmann fibers of the cerebellar cortex, tanycytes of the hypothalamus and ciliated ependymal cells) in the central nervous system (CNS).The antibody has recently been found to be a specific marker for low grade astrocytoma in human brain tissue. The antibody is able to distinguish between low grade astrocytoma and normal reactive gliosis (patent application filed). Monoclonal antibody J1-31 was raised against crude homogenate of brain tissue from a multiple sclerosis (MS) patient (autopsy sample; Malhotra et al.: Microbios Letters 26:151-157, 1984). In human brain, MAb J1-31 recognizes an intracellular protein antigen (J1-31 antigen), which bands at approximately 30,000 daltons under reducing conditions for sodium dodecyl sulfate gel electrophoresis (Singh et al.: Bioscience Reports 6:73-79, 1986). By immunofluorescence microscopy, MAb J1-31 stains those cells that are also stained by antiserum to glial fibrillary acidic protein (GFAP), namely astrocytes, retinal Muller cells, and tanycytes in the ependyma (Predy et al.: Bioscience Reports 7:491-502, 1987). In addition, MAb J1-31 stains ciliated ependymal cells that do not express GFAP (Malhotra, SK (1989) J Neurosci Res. 22(1):36-49).
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.