Learn More
Invitrogen™ ASXL1 Polyclonal Antibody

Description
Positive Control: human ASXL1-transfected 293T cells Predicted Reactivity: Mouse (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ASXL1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Asxl1 |
| Gene Accession No. | P59598, Q8IXJ9 |
| Gene Alias | additional sex combs like 1; additional sex combs like 1, transcriptional regulator; additional sex combs like transcriptional regulator 1; additional sex combs-like protein 1; Asxl1; BOPS; KIAA0978; MDS; MGC117280; MGC71111; mKIAA0978; Polycomb group protein ASXL1; putative Polycomb group protein ASXL1; RGD1561878 |
| Gene Symbols | Asxl1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.