Learn More
Tocris Bioscience™ AT 101 Inhibitor
Shop All R&D Systems Products

Description
Biological Activity:
R-(-)-enantiomer of gossypol. Mimics BH3 domains of Bcl-2, Bcl-XL and Mcl-1. Disrupts heterodimerization of Bcl-2 with proapoptotic family members. Induces apoptosis in vitro through activation of caspase-9; cytotoxic to multiple myeloma and drug-resistant cell lines. Delays onset of androgen-independent growth of VCaP prostate cancer xenografts in vivo.
Storage: Store the unopened product at 2° to 8°C. Do not use past expiration date.
Specifications
Specifications
| Formulation | C30H30O8 |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 10 mg |
| Product Type | Inhibits apoptosis |
| Molecular Weight (g/mol) | 518.56g/mol |
| Solubility | Soluble to 100mM in DMSO, 100mM in ethanol |
| Content And Storage | -20°C |
| Target | AT 101 |
| Color | Dark Purple |
For Laboratory Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.