Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AT 7867
GREENER_CHOICE

Catalog No. p-7235751
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent pan-Akt and PKA inhibitor

AT 7867 is a potent pan-Akt (protein kinase B) and protein kinase A (PKA) inhibitor (IC50 values are 32, 17, 47 and 20 nM for Akt1, Akt2, Akt3 and PKA, respectively). Also inhibits p70-S6 kinase (p70S6K, IC50 = 85 nM). Inhibits Akt signaling and proliferation of multiple human cancer cell lines, including breast, prostate and colon. Promotes proliferation of iPSC-derived PDX1+ pancreatic progenitor cells (PPCs).

Specifications

Chemical Name or Material 4-(4-Chlorophenyl)-4-[4-(1 H-pyrazol-4-yl)phenyl]piperidine hydrochloride
Quantity 10 mg
Target Akt (Protein Kinase B) Inhibitors
Molecular Formula C20H20ClN3.HCl
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.