Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATAD2 Antibody (CL0182), Novus Biologicals™
SDP

Catalog No. NB404922 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB404922 25ul
NBP214786 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB404922 Supplier Novus Biologicals Supplier No. NBP21478625UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATAD2 Monoclonal specifically detects ATAD2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen ATAD2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL0182
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 1:20, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 2-10 ug/mL, Immunohistochemistry-Paraffin 1:500-1:1000, Knockdown Validated
Gene Alias AAA nuclear coregulator cancer-associated protein, ANCCAMGC131938, ATPase family AAA domain-containing protein 2, ATPase family, AAA domain containing 2, DKFZp667N1320, EC 3.6.1.3, MGC142216, MGC29843, MGC5254, PRO2000
Gene Symbols ATAD2
Host Species Mouse
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: TAYAIIKEELDEDFEQLCEEIQESRKKRGCSSSKYAPSYYHVMPKQNSTLVGDKRSDPEQNEKLKTPSTPVACSTPAQLKRKIRKKSNWYLGTIKKRRKISQAKDDSQNAIDHK
Purification Method Protein A purified
Quantity 25ul
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 29028
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.