Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATAD5/FRAG1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140610
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9135 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9135 Supplier Bethyl Laboratories, Inc Supplier No. A301057AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. It has been found to be overexpressed in human lung adenocarcinomas and squamous cell carcinomas, and the overexpression of a fragment of the 3′UTR of the ZMYM4 mRNA termed Cell Death Inhibiting RNA (CDIR) protects HeLa cells from IFN-gamma-induced apoptosis, suggesting that ZMYM4 may play a role in tumorigenesis.

Specifications

Antigen ATAD5/FRAG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ATAD5
Gene Accession No. Q96QE3
Gene Alias C17orf41, chromosome 17 open reading frame 41, chromosome fragility associated gene 1, chromosome fragility-associated gene 1 protein, ELG1, enhanced level of genomic instability 1 homolog, enhanced level of genomic instability 1 homolog (S. cerevisiae), FLJ12735, FRAG1
Gene Symbols ATAD5
Host Species Rabbit
Immunogen Between 775 and 825
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79915
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.