Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ataxin 7 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291604 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291604 Supplier Novus Biologicals Supplier No. NBP142657APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ataxin 7 Polyclonal antibody specifically detects Ataxin 7 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen Ataxin 7
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias ADCAII, ataxin 7, FLJ17787, SCA7
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 842 and 892 of human spinocerebellar ataxia type 7 using the numbering given in entry NP_000324.1 (GeneID 6314).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6314
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.