Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ataxin 7-Like 3 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB326957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326957 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326957 Supplier Novus Biologicals Supplier No. NBP299131PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ataxin 7-Like 3 Polyclonal antibody specifically detects Ataxin 7-Like 3 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Ataxin 7-Like 3
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias Ataxin-7-Like Protein 3, ATXN7L3, SAGA-Associated Factor 11 Homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Ataxin 7-Like 3.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56970
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.