Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF1 Antibody (OTI2B4), PerCP, Novus Biologicals™
SDP

Catalog No. NB289736 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289736 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289736 Supplier Novus Biologicals Supplier No. NBP270218PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATF1 Monoclonal antibody specifically detects ATF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen ATF1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B4
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias activating transcription factor 1FUS/ATF-1, cAMP-dependent transcription factor ATF-1, EWS-ATF1, Protein TREB36, TREB36cyclic AMP-dependent transcription factor ATF-1
Host Species Mouse
Immunogen Full length human recombinant protein of human ATF1(NP_005162) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 466
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.